Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GN11 Mouse GnRH Neuronal Cell Line

Catalog No. SCC188MI Shop All MilliporeSigma Products
Change view
Click to view available options
Cell Type:
Neural Lineage Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type
SCC188MI Neural Lineage Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC188MI Supplier MilliporeSigma™ Supplier No. SCC188
Add to Cart
Add to Cart

GN11 mouse neuronal cell line is an immortalized cell line used as a model of GnRH-secreting neurons

  • Derived from transgenic mouse expressing Simian virus-40 (SV40) large T antigen driven by the human GnRH promoter
  • An olfactory bulb tumor from one male GnRH promoter-SV40T fusion-expressing transgenic mouse were dispersed and the GN11 cell line was established from serial dilution and cloning from primary cells
  • Uses:
    • Cell culturing
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in liquid nitrogen, Cells can be cultured for at least 10 passages after initial thawing without significantly affecting cell marker expression and functionality
    Cell Type Neural Lineage Cells
    For Use With (Application) Cell Culture, Model for GnRH-Secreting Neurons

    For research purposes only

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.