Learn More
Medchemexpress LLC HY-110266 5mg Medchemexpress, GN44028 CAS:1421448-26-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1102665MG
Encompass_Preferred
Medchemexpress, HY-110266 5mg GN44028 CAS:1421448-26-1 GN44028 is a hypoxia inducible factor (HIF)-1 inhibitor, with an IC50 of 14 nM. GN44028 inhibits hypoxia-induced HIF-1α transcriptional activity without suppressing HIF-1α mRNA expression, HIF-1α protein accumulation, or HIF-1α/HIF-1β heterodimerization. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8784