Learn More
Aviva Systems Biology Corp GNA11 Antibody - N-terminal region (ARP64960_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64960_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNA11. It was validated on Western Blot. . Alternate names: GNA-11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-401