Learn More
Aviva Systems Biology Corp GNAI2 Antibody - C-terminal region (ARP54632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNAI2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GNAI2. Alternate names: GIP, GNAI2B, H_LUCA15.1, H_LUCA16.1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61268