Learn More
Aviva Systems Biology Corp GNAI3 Antibody - N-terminal region (ARP58905_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58905_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNAI3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GNAI3. Alternate names: 87U6, FLJ26559ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-524