Learn More
ABclonal Technology GNAI3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A156741000UL
Encompass_Preferred
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling pathways. G proteins are composed of 3 units alpha, beta and gamma. This gene encodes an alpha subunit and belongs to the G-alpha family. Mutation in this gene, resulting in a gly40-to-arg substitution, is associated with auriculocondylar syndrome, and shown to affect downstream targets in the G protein-coupled endothelin receptor pathway.
Catalog No. 50-001-06499