Learn More
Aviva Systems Biology Corp GNAS Antibody - N-terminal region (ARP41764_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41764_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GNAS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GNAS. Alternate names: AHO, C20orf45, GNAS1, GPSA, GSA, GSP, MGC33735, PHP1A, PHP1B, POH, dJ309F20.1.1, dJ806M20.3.3, NESP, PHP1CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57987