Learn More
ABclonal Technology GNAZ Rabbit pAb
SDP
Supplier:  ABclonal Technology A147371000UL
Encompass_Preferred
The protein encoded by this gene is a member of a G protein subfamily that mediates signal transduction in pertussis toxin-insensitive systms. This encoded protein may play a role in maintaining the ionic balance of perilymphatic and endolymphatic cochlear fluids.
Catalog No. 50-001-08539