Learn More
Aviva Systems Biology Corp GNB1L Antibody - C-terminal region (ARP41328_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41328_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GNB1L. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GNB1L. Alternate names: GY2, FKSG1, WDR14, WDVCF, DGCRK3ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53294