Learn More
Aviva Systems Biology Corp RACK1 Antibody - N-terminal region (ARP61314_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61314_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gnb2l1. It was validated on Western Blot. . Alternate names: AL033335, GB-like, Gnb2-rs1, Rack1, p205ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-340