Learn More
Aviva Systems Biology Corp RACK1 Antibody - C-terminal region (ARP40625_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40625_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNB2L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GNB2L1. Alternate names: H12.3, HLC-7, PIG21, RACK1, Gnb2-rs1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-53216