Learn More
Aviva Systems Biology Corp RACK1 Antibody - N-terminal region (ARP40624_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40624_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNB2L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GNB2L1. Alternate names: Gnb2-rs1, H12.3, HLC-7, PIG21, RACK1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-57554