Learn More
Aviva Systems Biology Corp GNB5 Antibody - N-terminal region (ARP64092_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64092_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GNB5. It was validated on Western Blot. . Alternate names: FLJ37457, FLJ43714, GB5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-171