Learn More
Cayman Chemical Ganciclovir, Purity: ≥95%, CAS: 82410-32-0, 50 mg
SDP
Catalog No. 502954618
Encompass_Preferred
An active metabolite of valganciclovir and an antiviral nucleoside derivative; reduces human, monkey, mouse, and guinea pig CMV replication in plaque reduction assays (EC50s = 1.3-70 µM); topical application decreases disease severity and corneal leukocyte infiltration in a cat model of ocular FHV-1 epithelial infection at 0.15%; prevents allograft histologic injury and NK and myeloid cell infiltration in a mouse model of murine CMV infection with renal transplantation at 15 mg/kg; induces tumor regression in a K-BALB murine sarcoma model using a mixed population of HSV TK positive or negative cells
Catalog No. 50-295-4618 Supplier Cayman Chemical Supplier No. 1385350 MG
An active metabolite of valganciclovir and an antiviral nucleoside derivative; reduces human, monkey, mouse, and guinea pig CMV replication in plaque reduction assays (EC50s = 1.3-70 µM); topical application decreases disease severity and corneal leukocyte infiltration in a cat model of ocular FHV-1 epithelial infection at 0.15%; prevents allograft histologic injury and NK and myeloid cell infiltration in a mouse model of murine CMV infection with renal transplantation at 15 mg/kg; induces tumor regression in a K-BALB murine sarcoma model using a mixed population of HSV TK positive or negative cells