Learn More
Medchemexpress LLC HY-108435 10mg Medchemexpress, GNE-049 CAS:1936421-41-8 Purity:>98%
SDP
Catalog No. 501968547
Encompass_Preferred
Medchemexpress, HY-108435 10mg GNE-049 CAS:1936421-41-8 GNE-049 is a highly potent and selective CBP inhibitor with an IC50 of 1.1 nM in TR-FRET assay. GNE-049 also inhibits BRET and BRD4(1) with IC50s of 12 nM and 4200 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8547 Supplier Medchemexpress LLC Supplier No. HY10843510MG
Medchemexpress, HY-108435 10mg GNE-049 CAS:1936421-41-8 GNE-049 is a highly potent and selective CBP inhibitor with an IC50 of 1.1 nM in TR-FRET assay. GNE-049 also inhibits BRET and BRD4(1) with IC50s of 12 nM and 4200 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.