Learn More
Medchemexpress LLC GNE-049 | 1936421-41-8 | 99.7% | 510.58 | 25 MG
SDP
Catalog No. 5000347294
Encompass_Preferred
GNE-049 is a highly potent and selective CBP inhibitor. It demonstrates inhibition of BRET and BRD4(1).
  • Highly potent and selective CBP inhibitor
  • Inhibits BRET and BRD4(1)
  • Excellent potency in BRET cellular assay
  • Inhibits MYC expression
  • Acceptable pharmacokinetics in multiple species
  • Selective for CBP/P300 over BRD4(1)
Catalog No. 50-003-47294 Supplier Medchemexpress LLC Supplier No. 25MGHY108435
GNE-049 is a highly potent and selective CBP inhibitor. It demonstrates inhibition of BRET and BRD4(1).
  • Highly potent and selective CBP inhibitor
  • Inhibits BRET and BRD4(1)
  • Excellent potency in BRET cellular assay
  • Inhibits MYC expression
  • Acceptable pharmacokinetics in multiple species
  • Selective for CBP/P300 over BRD4(1)