Learn More
Abcam GNE-0877, 10MG
Catalog No. 502681663
Encompass_Preferred
MW 339.32 Da, Purity =98%. A potent and selective Leucine-rich repeat kinase 2 (LRRK2) inhibitor, Ki = 0.7 nM.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-1663 Supplier Abcam Supplier No. AB28244110MG
MW 339.32 Da, Purity =98%. A potent and selective Leucine-rich repeat kinase 2 (LRRK2) inhibitor, Ki = 0.7 nM.
The product is subject to the following: Abcam Restricted Use Statement