Learn More
Selleck Chemical LLC GNE-0877-5mg
SDP
Supplier:  Selleck Chemical LLC S73675MG
Encompass_Preferred
GNE-0877 (DNL201) is a highly potent and selective leucine-rich repeat kinase 2 (LRRK2) inhibitor with Ki of 0.7 nM.
Catalog No. 50-238-9708