Learn More
Medchemexpress LLC GNE-0946 | 1677667-24-1 | 98.1% | 10 MG
SDP
Catalog No. 5000294750
Encompass_Preferred
GNE-0946 is a potent, selective small-molecule agonist of the retinoic acid receptor-related orphan receptor γ (RORγ/RORc). It activates RORγ in HEK-293 cell-based assays with an EC50 of 4 nM. The compound is supplied as a research reagent for in vitro studies and is not intended for human or clinical use.
  • Potent RORγ agonist (EC50 4 nM in HEK-293 cells).
  • Selective activity suitable for ROR signaling studies.
  • High purity (~98.1%) appropriate for biochemical assays.
  • Chemical identity confirmed (CAS 1677667-24-1).
  • Small-molecule format compatible with cell-based and biochemical experiments.
  • Supplied as 10 mg for laboratory use.
Catalog No. 50-002-94750 Supplier Medchemexpress LLC Supplier No. HY1977410MG
GNE-0946 is a potent, selective small-molecule agonist of the retinoic acid receptor-related orphan receptor γ (RORγ/RORc). It activates RORγ in HEK-293 cell-based assays with an EC50 of 4 nM. The compound is supplied as a research reagent for in vitro studies and is not intended for human or clinical use.
  • Potent RORγ agonist (EC50 4 nM in HEK-293 cells).
  • Selective activity suitable for ROR signaling studies.
  • High purity (~98.1%) appropriate for biochemical assays.
  • Chemical identity confirmed (CAS 1677667-24-1).
  • Small-molecule format compatible with cell-based and biochemical experiments.
  • Supplied as 10 mg for laboratory use.