Learn More
Medchemexpress LLC GNE-131 | 1629063-81-5 | 98.2% | 442.57 | 1 ML
SDP
Catalog No. 5000438177
Encompass_Preferred
GNE-131 is a potent and selective inhibitor of human sodium channels, specifically targeting NaV1.7 with an IC50 of 3 nM. This compound has demonstrated excellent potency and good in vitro metabolic stability. It also exhibits low in vivo clearance in mouse, rat, and dog models, along with excellent efficacy in a transgenic mouse model of induced pain.
  • Potent and selective sodium channel inhibition.
  • Good metabolic stability in laboratory settings.
  • Effective in pain models.
  • Low clearance in multiple animal species.
Catalog No. 50-004-38177 Supplier Medchemexpress LLC Supplier No. HY11227910MM/1ML
GNE-131 is a potent and selective inhibitor of human sodium channels, specifically targeting NaV1.7 with an IC50 of 3 nM. This compound has demonstrated excellent potency and good in vitro metabolic stability. It also exhibits low in vivo clearance in mouse, rat, and dog models, along with excellent efficacy in a transgenic mouse model of induced pain.
  • Potent and selective sodium channel inhibition.
  • Good metabolic stability in laboratory settings.
  • Effective in pain models.
  • Low clearance in multiple animal species.