Learn More
Selleck Chemical LLC -GNE-140 2mg
SDP
Supplier:  Selleck Chemical LLC S66752MG
Encompass_Preferred
(R)-GNE-140 is a selective inhibitor of the LDHA and LDHB with IC50s of 3 nM and 5 nM, respectively. The R enantiomer of GNE-140 is 18-fold more potent than S enantiomer. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3205