Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GNE-149 | 1953132-75-6 | 99.2% | C28H33F4N3O | 1 ML
SDP

Catalog No. 5000355229
Encompass_Preferred

GNE-149 is an orally bioavailable full antagonist of estrogen receptor α (ERα; IC50=0.053 nM). It is a selective estrogen receptor degrader (SERD) and can be used for the research of breast cancer.

  • Appearance: solid
  • Color: white to light yellow
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-003-55229 Supplier Medchemexpress LLC Supplier No. HY14534110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GNE-149 is an orally bioavailable full antagonist of estrogen receptor α (ERα; IC50=0.053 nM). It is a selective estrogen receptor degrader (SERD) and can be used for the research of breast cancer.

  • Appearance: solid
  • Color: white to light yellow
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.