Learn More
Medchemexpress LLC HY-120028 5mg Medchemexpress, GNE-207 CAS:2158266-58-9 Purity:>98%
SDP
Catalog No. 501969569
Encompass_Preferred
Medchemexpress, HY-120028 5mg GNE-207 CAS:2158266-58-9 GNE-207 is a potent, selective and orally bioavailable inhibitor of the bromodomain of CBP, with an IC50 of 1 nM, exhibits a selectively index of >2500-fold against BRD4 (1). GNE-207 shows excellent CBP potency, with an EC50 of 18 nM for MYC expression in MV-4-11 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9569 Supplier Medchemexpress LLC Supplier No. HY1200285MG
Medchemexpress, HY-120028 5mg GNE-207 CAS:2158266-58-9 GNE-207 is a potent, selective and orally bioavailable inhibitor of the bromodomain of CBP, with an IC50 of 1 nM, exhibits a selectively index of >2500-fold against BRD4 (1). GNE-207 shows excellent CBP potency, with an EC50 of 18 nM for MYC expression in MV-4-11 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.