Learn More
Medchemexpress LLC GNE-235 | 3036574-72-5 | C13H16N4O | 5 MG
SDP
Catalog No. 5000375574
Encompass_Preferred
GNE-235 is a compound selective for the second bromodomain of PBRM1, with a KD of 0.28 ± 0.02 μM. It can be used for evaluation of the cellular function of PBRM1.
  • Selective for the second bromodomain of PBRM1
  • Utilized for assessing the cellular function of PBRM1
  • For research use only
  • Epigenetic reader domain inhibitor
  • Related to cancer, blood or cardio-cerebrovascular disease, and leukemia/lymphoma/myeloma
Catalog No. 50-003-75574 Supplier Medchemexpress LLC Supplier No. HY1502895MG
GNE-235 is a compound selective for the second bromodomain of PBRM1, with a KD of 0.28 ± 0.02 μM. It can be used for evaluation of the cellular function of PBRM1.
  • Selective for the second bromodomain of PBRM1
  • Utilized for assessing the cellular function of PBRM1
  • For research use only
  • Epigenetic reader domain inhibitor
  • Related to cancer, blood or cardio-cerebrovascular disease, and leukemia/lymphoma/myeloma