Learn More
Medchemexpress LLC GNE 2861 | 1394121-05-1 | 98.97% | 406.48 | 100 MG
SDP
Catalog No. 5000364858
Encompass_Preferred
GNE 2861 is a PAK inhibitor that exhibits group II selectivity. It inhibits PAK4, PAK5, and PAK6 with IC50 values of 7.5 nM, 36 nM, and 126 nM, respectively.
  • PAK inhibitor
  • Exhibits group II selectivity
  • Inhibits PAK4 with an IC50 of 7.5 nM
  • Inhibits PAK5 with an IC50 of 36 nM
  • Inhibits PAK6 with an IC50 of 126 nM
Catalog No. 50-003-64858 Supplier Medchemexpress LLC Supplier No. 100MGHY12632
GNE 2861 is a PAK inhibitor that exhibits group II selectivity. It inhibits PAK4, PAK5, and PAK6 with IC50 values of 7.5 nM, 36 nM, and 126 nM, respectively.
  • PAK inhibitor
  • Exhibits group II selectivity
  • Inhibits PAK4 with an IC50 of 7.5 nM
  • Inhibits PAK5 with an IC50 of 36 nM
  • Inhibits PAK6 with an IC50 of 126 nM