Learn More
Apexbio Technology LLC GNE-317, 50mg Cas# 1394076-92-6
SDP
Supplier:  Apexbio Technology LLC B499150
Encompass_Preferred
GNE-317 5mg. 214358-33-5. MFCD03428622 GNE-317 is a potent inhibitor of PI3K [1]. Phosphatidylinositol-4,5-bisphosphate 3-kinase (PI3K) are a series of enzymes and play an important role in cell growth, proliferation, differentiation, survival, motility and intracellular trafficking. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8710