Learn More
Medchemexpress LLC GNE-371 | 1926986-36-8 | 98.1% | 431.49 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11280350MG
Encompass_Preferred
GNE-371 is a potent and selective chemical probe designed for the second bromodomains of human transcription-initiation-factor TFIID subunit 1 and transcription-initiation-factor TFIID subunit 1-like. It demonstrates an IC50 of 10 nM for TAF1(2).
  • Potent and selective chemical probe for TAF1(2).
  • Binds TAF1(2) with an IC50 of 10 nM.
  • Maintains excellent selectivity over other bromodomain-family members.
  • Active in a cellular-TAF1(2) target-engagement assay (IC50=38 nM).
  • Exhibits anti-proliferative synergy with the BET inhibitor JQ1.
  • Useful for mechanistic and target-validation studies.
Catalog No. 50-004-37387