Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gne-371 | 1926986-36-8 | 98.2% | 431.5 g/mol | C24H25N5O3 | 5 MG
SDP

Catalog No. 5000262488
Encompass_Preferred

GNE-371 is a potent, selective chemical probe that targets the second bromodomain of human transcription-initiation factor TFIID subunit 1 (TAF1). It is provided as a research-grade small molecule for biochemical and cellular assays with reported low-nanomolar activity against TAF1(2).

  • Potent, selective TAF1(2) bromodomain inhibitor with IC50 ≈ 10 nM.
  • Suitable as a chemical probe for biochemical and cellular studies.
  • High purity (98.2%).
  • Molecular formula C24H25N5O3.
  • Molecular weight 431.5 g/mol.
  • CAS number 1926986-36-8.
  • Soluble in DMSO for assay preparation.

Catalog No. 50-002-62488 Supplier Medchemexpress LLC Supplier No. HY1128035MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GNE-371 is a potent, selective chemical probe that targets the second bromodomain of human transcription-initiation factor TFIID subunit 1 (TAF1). It is provided as a research-grade small molecule for biochemical and cellular assays with reported low-nanomolar activity against TAF1(2).

  • Potent, selective TAF1(2) bromodomain inhibitor with IC50 ≈ 10 nM.
  • Suitable as a chemical probe for biochemical and cellular studies.
  • High purity (98.2%).
  • Molecular formula C24H25N5O3.
  • Molecular weight 431.5 g/mol.
  • CAS number 1926986-36-8.
  • Soluble in DMSO for assay preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.