Learn More
Selleck Chemical LLC GNE-477
SDP
Catalog No. 502173182
Encompass_Preferred
GNE-477 is a potent and efficacious dual PI3K/mTOR inhibitor with an IC50 of 4 nM for PI3K and Kiapp of 21 nM for mTOR
Catalog No. 50-217-3182 Supplier Selleck Chemical LLC Supplier No. S65161MG
GNE-477 is a potent and efficacious dual PI3K/mTOR inhibitor with an IC50 of 4 nM for PI3K and Kiapp of 21 nM for mTOR