Learn More
Medchemexpress LLC HY-11042 5mg Medchemexpress, GNE-477 CAS:1032754-81-6 Purity:>98%
SDP
Catalog No. 501968798
Encompass_Preferred
Medchemexpress, HY-11042 5mg GNE-477 CAS:1032754-81-6 GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8798 Supplier Medchemexpress LLC Supplier No. HY110425MG
Medchemexpress, HY-11042 5mg GNE-477 CAS:1032754-81-6 GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.