Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gne-493 | 1033735-94-2 | MFCD18251526 | 99.8% | 372.44 g/mol | C17H20N6O2S | 1 ML
SDP

Catalog No. 5000242461
Encompass_Preferred

GNE-493 is a potent, selective, orally available small-molecule inhibitor of class I PI3K isoforms and mTOR, with demonstrated biochemical potency and cellular antiproliferative activity. It is supplied as a solid or as ready-to-use DMSO solutions (for example, 10 mM, 1 mL).

  • Dual pan-PI3K and mTOR inhibition with low-nanomolar IC50s.
  • Biochemical IC50s: PI3Kα 3.4 nM; PI3Kβ 12 nM; PI3Kδ 16 nM; PI3Kγ 16 nM; mTOR 30 nM.
  • Cellular antiproliferative activity (PC-3 IC50 0.33 μM).
  • High purity and defined physicochemical profile (MW 372.44; formula C17H20N6O2S).
  • Available as solution in DMSO or as a solid for custom formulation.

Catalog No. 50-002-42461 Supplier Medchemexpress LLC Supplier No. HY1081110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GNE-493 is a potent, selective, orally available small-molecule inhibitor of class I PI3K isoforms and mTOR, with demonstrated biochemical potency and cellular antiproliferative activity. It is supplied as a solid or as ready-to-use DMSO solutions (for example, 10 mM, 1 mL).

  • Dual pan-PI3K and mTOR inhibition with low-nanomolar IC50s.
  • Biochemical IC50s: PI3Kα 3.4 nM; PI3Kβ 12 nM; PI3Kδ 16 nM; PI3Kγ 16 nM; mTOR 30 nM.
  • Cellular antiproliferative activity (PC-3 IC50 0.33 μM).
  • High purity and defined physicochemical profile (MW 372.44; formula C17H20N6O2S).
  • Available as solution in DMSO or as a solid for custom formulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.