Learn More
Medchemexpress LLC HY-10811 10mg Medchemexpress, GNE-493 CAS:1033735-94-2 Purity:>98%
SDP
Catalog No. 501968508
Encompass_Preferred
Medchemexpress, HY-10811 10mg GNE-493 CAS:1033735-94-2 GNE-493 is a potent, selective, and orally available dual pan-PI3-kinase/mTOR inhibitor with IC50s of 3.4 nM, 12 nM, 16 nM, 16 nM and 32 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8508 Supplier Medchemexpress LLC Supplier No. HY1081110MG
Medchemexpress, HY-10811 10mg GNE-493 CAS:1033735-94-2 GNE-493 is a potent, selective, and orally available dual pan-PI3-kinase/mTOR inhibitor with IC50s of 3.4 nM, 12 nM, 16 nM, 16 nM and 32 nM for PI3Kα, PI3Kβ, PI3Kδ, PI3Kγ and mTOR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.