Learn More
Medchemexpress LLC HY-100343 1mg , GNE-495 CAS:1449277-10-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1003431MG
Encompass_Preferred
HY-100343 1mg GNE-495 CAS: 1449277-10-4 GNE-495 is a potent and selective MAP4K4 inhibitor with an IC50 of 3.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0358