Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GNE 5729 | 2026635-66-3 | MFCD30829916 | 99.1% | 428.20 g·mol⁻¹ | C17H10Cl2F3N5O | 50 MG
SDP

Catalog No. 5000241378
Encompass_Preferred

GNE 5729 is a brain-permeable positive allosteric modulator of the N-methyl-D-aspartate receptor (NMDAR) used for biochemical and pharmacological research. It displays subtype-selective activity and physicochemical properties suitable for in vitro and in vivo studies.

  • Brain-permeable positive allosteric modulator of NMDAR.
  • Potent activity at GluN2A (EC50 37 nM); micromolar activity at GluN2C and GluN2D.
  • High purity reported at 99.05%.
  • Molecular weight 428.20 g·mol⁻¹ and formula C17H10Cl2F3N5O.
  • Soluble in DMSO for experimental preparations.
  • Available in small research quantities such as 50 MG.

Catalog No. 50-002-41378 Supplier Medchemexpress LLC Supplier No. HY10740950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GNE 5729 is a brain-permeable positive allosteric modulator of the N-methyl-D-aspartate receptor (NMDAR) used for biochemical and pharmacological research. It displays subtype-selective activity and physicochemical properties suitable for in vitro and in vivo studies.

  • Brain-permeable positive allosteric modulator of NMDAR.
  • Potent activity at GluN2A (EC50 37 nM); micromolar activity at GluN2C and GluN2D.
  • High purity reported at 99.05%.
  • Molecular weight 428.20 g·mol⁻¹ and formula C17H10Cl2F3N5O.
  • Soluble in DMSO for experimental preparations.
  • Available in small research quantities such as 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.