Learn More
Medchemexpress LLC HY-112937 10mg Medchemexpress, GNE-6640 CAS:2009273-67-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11293710MG
Encompass_Preferred
Medchemexpress, HY-112937 10mg GNE-6640 CAS:2009273-67-8 GNE-6640 is a selective and non-covalent inhibitor of ubiquitin epecific peptidase 7 (USP7), with IC50 values of 0.75 μM, 0.43 μM, 20.3 μM and 0.23 μM for full length USP7, USP7 catalytic domain, full length USP43 and Ub-MDM2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9210