Learn More
Medchemexpress LLC HY-108696 5mg Medchemexpress, GNE-781 CAS:1936422-33-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1086965MG
Encompass_Preferred
Medchemexpress, HY-108696 5mg GNE-781 CAS:1936422-33-1 GNE-781 is a highly potent and selective CBP inhibitor with an IC 50 of 0.94 nM in TR-FRET assay. GNE-781 also inhibits BRET and BRD4(1) with IC 50 s of 6.2 nM and 5100 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2329