Learn More
Medchemexpress LLC GNE-7883 | 2648450-42-2 | 99.5% | 500.57 | 100 MG
SDP
Catalog No. 5000362859
Encompass_Preferred
GNE-7883 is a pan-TEAD inhibitor that blocks the association of YAP/TAZ with TEAD. It effectively reduces chromatin accessibility at TEAD motifs, inhibits cell proliferation in multiple cell line models, and shows strong anti-tumor efficacy in vivo. Additionally, GNE-7883 overcomes intrinsic and acquired resistance to KRAS G12C inhibitors.
  • Pan-TEAD inhibitor
  • Blocks YAP/TAZ association with TEAD
  • Reduces chromatin accessibility at TEAD motifs
  • Inhibits cell proliferation
  • Shows anti-tumor efficacy in vivo
  • Overcomes KRAS G12C inhibitor resistance
  • Targets YAP/TAZ-TEAD
  • Soluble in DMSO (50 mg/mL)
Catalog No. 50-003-62859 Supplier Medchemexpress LLC Supplier No. HY147214100MG
GNE-7883 is a pan-TEAD inhibitor that blocks the association of YAP/TAZ with TEAD. It effectively reduces chromatin accessibility at TEAD motifs, inhibits cell proliferation in multiple cell line models, and shows strong anti-tumor efficacy in vivo. Additionally, GNE-7883 overcomes intrinsic and acquired resistance to KRAS G12C inhibitors.
  • Pan-TEAD inhibitor
  • Blocks YAP/TAZ association with TEAD
  • Reduces chromatin accessibility at TEAD motifs
  • Inhibits cell proliferation
  • Shows anti-tumor efficacy in vivo
  • Overcomes KRAS G12C inhibitor resistance
  • Targets YAP/TAZ-TEAD
  • Soluble in DMSO (50 mg/mL)