Learn More
Medchemexpress LLC HY-18163A 5mg Medchemexpress, GNE-7915 (tosylate) CAS:2070015-00-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18163A5MG
Encompass_Preferred
Medchemexpress, HY-18163A 5mg GNE-7915 (tosylate) CAS:2070015-00-6 GNE-7915 tosylate is a potent, selective and brain-penetrant inhibitor of LRRK2 with an IC50 of 9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2022