Learn More
Medchemexpress LLC GNE-886 | 2101957-05-3 | 99.8% | 498.58 | C28H30N6O3 | 50 MG
SDP
Catalog No. 5000226546
Encompass_Preferred
GNE-886 is a potent, selective small-molecule inhibitor of the CECR2 bromodomain used for epigenetic reader-domain research and cellular assays. Supplied as a 50 mg research sample with reported purity 99.81%, it has molecular formula C28H30N6O3 and molecular weight 498.58; reported potency is IC50 = 0.016 μM (CECR2) and EC50 = 370 nM in cellular assays.
  • Potent and selective CECR2 bromodomain inhibition.
  • High purity (99.81%) suitable for biochemical and cellular studies.
  • Small-scale package for research use and lead optimization.
  • Well-characterized by molecular formula and molecular weight.
  • Datasheet and certificate of analysis available.
Catalog No. 50-002-26546 Supplier Medchemexpress LLC Supplier No. HY11786550MG
GNE-886 is a potent, selective small-molecule inhibitor of the CECR2 bromodomain used for epigenetic reader-domain research and cellular assays. Supplied as a 50 mg research sample with reported purity 99.81%, it has molecular formula C28H30N6O3 and molecular weight 498.58; reported potency is IC50 = 0.016 μM (CECR2) and EC50 = 370 nM in cellular assays.
  • Potent and selective CECR2 bromodomain inhibition.
  • High purity (99.81%) suitable for biochemical and cellular studies.
  • Small-scale package for research use and lead optimization.
  • Well-characterized by molecular formula and molecular weight.
  • Datasheet and certificate of analysis available.