Learn More
Medchemexpress LLC GNE-987 GSH linker-1 | 2417370-39-7 | C62H77F2N9O12S4 | 10MG
SDP
Catalog No. 5000260649
Encompass_Preferred
GNE-987 GSH linker-1 is a PROTAC-linker conjugate that pairs a BET-degrading warhead with a disulfide-containing glutathione-cleavable linker for use in PROTAC-antibody conjugate (PAC) research. It is intended as a research reagent for chemical biology studies involving targeted protein degradation and intracellular payload release.
  • Enables targeted protein degradation when conjugated to antibodies.
  • Contains a disulfide-based glutathione-cleavable linker for intracellular release.
  • High chemical purity suitable for analytical and biological testing.
  • Compatible with common solvents and recommended storage protocols.
  • Applicable to studies of protein degradation pathways and PAC development.
Catalog No. 50-002-60649 Supplier Medchemexpress LLC Supplier No. HY12993810MG
GNE-987 GSH linker-1 is a PROTAC-linker conjugate that pairs a BET-degrading warhead with a disulfide-containing glutathione-cleavable linker for use in PROTAC-antibody conjugate (PAC) research. It is intended as a research reagent for chemical biology studies involving targeted protein degradation and intracellular payload release.
  • Enables targeted protein degradation when conjugated to antibodies.
  • Contains a disulfide-based glutathione-cleavable linker for intracellular release.
  • High chemical purity suitable for analytical and biological testing.
  • Compatible with common solvents and recommended storage protocols.
  • Applicable to studies of protein degradation pathways and PAC development.