Learn More
Apexbio Technology LLC GNF 2 778270-11-4 5mg
SDP
Catalog No. 502240502
Encompass_Preferred
GNF 2 (CAS 778270-11-4) is a small-molecule inhibitor targeting Bcr-Abl kinase It is designed to suppress Bcr-Abl-mediated signaling thereby regulating leukemic cell proliferation GNF 2 exerts its biological activity primarily through non-ATP competitive allosteric inhibition at the myristate-binding pocket of Abl In cell-based studies GNF 2 demonstrates inhibitory activity with an IC50 of approximately 138 nM in Ba/F3 p210 cells expressing wild-type Bcr-Abl It also inhibits mutant Bcr-Abl forms (E255V Y253H) and proliferation of K562 and SUP-B15 human leukemia cell lines with IC50 values of 194 273 nM Based on these pharmacological properties GNF 2 holds research potential in chronic myeloid leukemia (CML) pathogenesis and therapeutic response studies involving Bcr-Abl fusion proteins
This item is not returnable. View return policy
Catalog No. 50-224-0502 Supplier Apexbio Technology LLC Supplier No. A86035
GNF 2 (CAS 778270-11-4) is a small-molecule inhibitor targeting Bcr-Abl kinase It is designed to suppress Bcr-Abl-mediated signaling thereby regulating leukemic cell proliferation GNF 2 exerts its biological activity primarily through non-ATP competitive allosteric inhibition at the myristate-binding pocket of Abl In cell-based studies GNF 2 demonstrates inhibitory activity with an IC50 of approximately 138 nM in Ba/F3 p210 cells expressing wild-type Bcr-Abl It also inhibits mutant Bcr-Abl forms (E255V Y253H) and proliferation of K562 and SUP-B15 human leukemia cell lines with IC50 values of 194 273 nM Based on these pharmacological properties GNF 2 holds research potential in chronic myeloid leukemia (CML) pathogenesis and therapeutic response studies involving Bcr-Abl fusion proteins