Learn More
TARGETMOL CHEMICALS INC GNF-5 50MG
SDP
Catalog No. 502577298
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GNF-5 is a specific non-ATP competitive inhibitor of Bcr-Abl (IC50 0.22±0.1 uM Wild-type Abl). It is an analog of GNF-2 with improved pharmacokinetic properties. purity: 99%
Catalog No. 50-257-7298 Supplier TARGETMOL CHEMICALS INC Supplier No. T180350MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GNF-5 is a specific non-ATP competitive inhibitor of Bcr-Abl (IC50 0.22±0.1 uM Wild-type Abl). It is an analog of GNF-2 with improved pharmacokinetic properties. purity: 99%