Learn More
TARGETMOL CHEMICALS INC GNF-7 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T319610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GNF-7 is Bcr-Abl WT and Bcr-Abl T315I inhibitor with IC50 of 133 nM and 61 nM respectively. purity: 99%
Catalog No. 50-258-0341