Learn More
TARGETMOL CHEMICALS INC GNF-8625 monopyridin-N-piperaz
SDP
Supplier:  TARGETMOL CHEMICALS INC T400345MG
Encompass_Preferred
Also available in 1 mg, 10 mg and bulk. Please contact Fisher for quotes. GNF-8625 monopyridin-N-piperazine hydrochloride is a tropomyosin receptor kinase (TRK) inhibitor. Purity 99.44%
Catalog No. 50-287-8987