Learn More
Medchemexpress LLC HY-100687 5mg Medchemexpress, GNF-PF-3777 CAS:77603-42-0 Purity:>98%
SDP
Catalog No. 501931915
Encompass_Preferred
Medchemexpress, HY-100687 5mg GNF-PF-3777 CAS:77603-42-0 GNF-PF-3777 (8-Nitrotryptanthrin) is a potent human indoleamine 2,3-dioxygenase 2 (h IDO2 ) inhibitor which significantly reduces IDO2 activity with K i of 0.97 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1915 Supplier Medchemexpress LLC Supplier No. HY1006875MG
Medchemexpress, HY-100687 5mg GNF-PF-3777 CAS:77603-42-0 GNF-PF-3777 (8-Nitrotryptanthrin) is a potent human indoleamine 2,3-dioxygenase 2 (h IDO2 ) inhibitor which significantly reduces IDO2 activity with K i of 0.97 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.