Learn More
Medchemexpress LLC HY-142295 5mg , GNF2133 CAS:2561414-56-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1422955MG
Encompass_Preferred
HY-142295 5mg GNF2133 CAS: 2561414-56-8 GNF2133 is a potent, selective and orally active DYRK1A inhibitor with IC50s of 0.0062, >50 µM for DYRK1A and GSK3β, respectively. GNF2133 shows good proliferation potency and efficacy on rat and human primary β-cell. GNF2133 significantly improves glucose disposal capacity and increases insulin secretion. GNF2133 has the potential for the research of type 1 diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1847