Learn More
Medchemexpress LLC HY-102023 10mg Medchemexpress, GNF351 CAS:1227634-69-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10202310MG
Encompass_Preferred
Medchemexpress, HY-102023 10mg GNF351 CAS:1227634-69-6 GNF351 is a full aryl hydrocarbon receptor (AHR) antagonist. GNF351 competes with a photoaffinity AHR ligand for binding to the AHR with an IC50 of 62 nM. GNF351 is minimal toxicity in mouse or human keratinocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7953