Learn More
Cayman Chemical Gingerenone A, Purity: ≥95%, CAS: 128700-97-0, 10 mg
SDP
Catalog No. 502909325
Encompass_Preferred
A diarylheptanoid and polyphenol with diverse biological activities; inhibits tyrosinase (IC50 = 9.4 µM for the mushroom enzyme); scavenges DPPH radicals in a cell-free assay (IC50 = 12.6 µM); inhibits JAK2 and p70S6K1 at 5 and 10 µM; selectively decreases the viability of ionizing radiation-induced senescent WI38 fibroblasts over proliferating fibroblasts at 20 µM; prevents increases in body weight but does not affect caloric intake in a mouse model of high-fat diet-induced obesity at 50 mg/kg; decreases tumor weight in an HCT116 colon cancer mouse xenograft model at 5 and 20 mg/kg
Catalog No. 50-290-9325 Supplier Cayman Chemical Supplier No. 3684110 MG
A diarylheptanoid and polyphenol with diverse biological activities; inhibits tyrosinase (IC50 = 9.4 µM for the mushroom enzyme); scavenges DPPH radicals in a cell-free assay (IC50 = 12.6 µM); inhibits JAK2 and p70S6K1 at 5 and 10 µM; selectively decreases the viability of ionizing radiation-induced senescent WI38 fibroblasts over proliferating fibroblasts at 20 µM; prevents increases in body weight but does not affect caloric intake in a mouse model of high-fat diet-induced obesity at 50 mg/kg; decreases tumor weight in an HCT116 colon cancer mouse xenograft model at 5 and 20 mg/kg