Learn More
Cayman Chemical GnglIosID GM1 MoncLNal n 100ug
SDP
Catalog No. 502916701
Encompass_Preferred
Immunogen: C. jejuni OH4384 • Host: Mouse • Cross Reactivity: (+) Ganglioside GM1; (-) Ganglioside GM2, Ganglioside GM3, Ganglioside GD1a, Ganglioside GD1B, Ganglioside GD3 • Applications: Dot blot, ELISA, IHC
Catalog No. 50-291-6701 Supplier Cayman Chemical Supplier No. 38289100 UG
Immunogen: C. jejuni OH4384 • Host: Mouse • Cross Reactivity: (+) Ganglioside GM1; (-) Ganglioside GM2, Ganglioside GM3, Ganglioside GD1a, Ganglioside GD1B, Ganglioside GD3 • Applications: Dot blot, ELISA, IHC