Learn More
Cayman Chemical Ganglioside GM1 (bovine) (sodium salt), Purity: ≥98%, CAS: 37758-47-7, 5 mg
SDP
Catalog No. 502944814
Encompass_Preferred
A mixture of ganglioside GM1 species isolated from bovine brain with primarily C18:0 acyl chain lengths; interacts with other proteins to increase calcium influx, affecting various calcium-dependent processes, including inducing neuronal outgrowth during differentiation; acts as a receptor for cholera toxin and the heat-labile enterotoxin from E. coli; induces apoptosis in activated T cells; found at higher levels on T cells isolated from patients with RCC; levels are decreased in the substantia nigra pars compacta in postmortem brain from patients with Parkinson's disease; accumulates in neurons in ganglioside GM1 gangliosidosis
Catalog No. 50-294-4814 Supplier Cayman Chemical Supplier No. 412065 MG
A mixture of ganglioside GM1 species isolated from bovine brain with primarily C18:0 acyl chain lengths; interacts with other proteins to increase calcium influx, affecting various calcium-dependent processes, including inducing neuronal outgrowth during differentiation; acts as a receptor for cholera toxin and the heat-labile enterotoxin from E. coli; induces apoptosis in activated T cells; found at higher levels on T cells isolated from patients with RCC; levels are decreased in the substantia nigra pars compacta in postmortem brain from patients with Parkinson's disease; accumulates in neurons in ganglioside GM1 gangliosidosis